Impurity and API standards, Diethyl (4RS)-2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Catalog # PROOMM0383.10
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standards
- CAS No.:140171-65-9
- Description:Diethyl (4RS)-2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
- Formula:C₂₁H₂₇ClN₂O₅
- Description:Diethyl (4RS)-2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate