Order Entry
Belgium
ContactUsLinkComponent
Impurity and API standards, (1RS,4RS)-4-(4-Chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride
Impurity and API standards, (1RS,4RS)-4-(4-Chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride
Catalog # PROOMM0317.06
CAS Number:  
Impurity and API standards, (1RS,4RS)-4-(4-Chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride
Catalog # PROOMM0317.06
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    79559-98-1
  • Description:
    (1RS,4RS)-4-(4-Chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride
  • Formula:
    C₁₇H₁₈ClN·ClH
  • Description:
    (1RS,4RS)-4-(4-Chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

Specifications

About this item

Sertraline impurity: Imp. C (EP) as hydrochloride