Order Entry
Belgium
ContactUsLinkComponent
Impurity and API standard, 2-amino-7-[(2-hydroxyethoxy)methyl]-1,7-dihydro-6H-purin-6-one
Impurity and API standard, 2-amino-7-[(2-hydroxyethoxy)methyl]-1,7-dihydro-6H-purin-6-one
Catalog # PROOMM0061.03
CAS Number:  
Impurity and API standard, 2-amino-7-[(2-hydroxyethoxy)methyl]-1,7-dihydro-6H-purin-6-one
Catalog # PROOMM0061.03
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    91702-61-3
  • Description:
    2-amino-7-[(2-hydroxyethoxy)methyl]-1,7-dihydro-6H-purin-6-one
  • Formula:
    C₈H₁₁N₅O₃
  • Description:
    2-Amino-7-[(2-hydroxyethoxy)methyl]-1,7-dihydro-6H-purin-6-one

Specifications

About this item