Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, plastic additives, Ph. Eur., RS
Organic reference standards, plastic additives, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, plastic additives, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    301-02-0
  • Formula:
    C₁₈H₃₅NO
  • Description:
    Oleamide R

Specifications

Frequently Bought Together