Organic reference standards, pentoxyverine, Ph. Eur., RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:77-22-5
- Formula:C₁₈H₂₇NO₂
- Description:Pentoxyverine impurity B, 2-(diethylamino)ethyl 1-phenylcyclopentanecarboxylate (caramiphen)
Specifications
Frequently Bought Together