Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, norfloxacin, Ph. Eur., RS
Organic reference standards, norfloxacin, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, norfloxacin, Ph. Eur., RS
Supplier:  EDQM
Certificates

Specifications

  • CAS No.:
    70458-96-7
  • Formula:
    C₁₆H₁₈FN₃O₃
  • Description:
    Norfloxacin for peak identification

Specifications

Related Information

Frequently Bought Together