ContactUsLinkComponent
VWR®, Silane Coated, Double Frosted Slides
VWR®, Silane Coated, Double Frosted Slides
Catalog # 631-1162
Supplier:  VWR Collection
VWR®, Silane Coated, Double Frosted Slides
Catalog # 631-1162
Supplier:  VWR Collection
Specifications
  • W×D:
    76×25 mm
  • Description:
    Double-frosted silane coated slides
  • Seq:
    007
  • Thickness:
    1,0 (-0,05/+0,1) mm
  • Pk:
    2.500
Specifications
About this item
Silane (aminoalkylsilane) coating provide permanent adhesion, which can bind tissue sections to glass slides with a sticky surface.
  • Smooth edges for operational safety
  • Marking with pencils, marker pens and conventional labelling systems
  • Shelf life of 18 months
Recommended for H&E stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: Fragile items should be stored in dry and proper laboratory temperature conditions.