Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Plain Microscope Slides

VWR®, Plain Microscope Slides

Ground edges, clipped corner (45°) slides, 1,0 (-0,05/+0,1) mm, 76×26 mm
Catalog # 631-1160
Supplier:   VWR Collection
VWR®, Plain Microscope Slides
Catalog # 631-1160
Supplier:  VWR Collection
VWR®, Plain Microscope Slides

Ground edges, clipped corner (45°) slides, 1,0 (-0,05/+0,1) mm, 76×26 mm

Price
131,00 €
Log in for availability
1 * 2.500 ST
Product Details & Documents

Microscope slides with ground edges designed for human blood or fluid tests.

  • Shelf life of 60 months
  • Smooth edges for operational safety

Suitable for use in haematology and microbiology applications.

Packaging: 50 pieces per box. 50 boxes per case.

Caution: Fragile items should be stored in dry and proper laboratory temperature conditions.

Specifications
  • Description:
    Ground edges, clipped corner (45°) slides
  • L×W:
    76×26 mm
  • Thickness:
    1,0 (-0,05/+0,1) mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.