Orders LinkContactUsLinkComponent
Rods, ceramic, round, solid, AL23
Rods, ceramic, round, solid, AL23
Rods, ceramic, round, solid, AL23
Certificates
Rods, ceramic, round, solid, AL23
6 Options Available Below
Product Details & Documents
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Product Family Options
0,6 mm Ø
Product Information
  • Ext.Ø
    Length
    Availability
    Price
  • 0,6 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 100 mm
    1 * 100 mm
5,0 mm Ø
Product Information
  • Ext.Ø
    Length
    Availability
    Price
  • 5,0 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 200 mm
    1 * 200 mm - 61,20 €
    61,20 €
    1 * 200 mm
       61,20 €
  • 5,0 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 250 mm
    1 * 250 mm - 77,80 €
    77,80 €
    1 * 250 mm
       77,80 €
  • 5,0 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 300 mm
    1 * 300 mm - 80,40 €
    80,40 €
    1 * 300 mm
       80,40 €
  • 5,0 mm
    600 mm
    Log in for availability
    Log in for availability
    1 * 600 mm
    1 * 600 mm - 138,00 €
    138,00 €
    1 * 600 mm
       138,00 €
6,0 mm Ø
Product Information
  • Ext.Ø
    Length
    Availability
    Price
  • 6,0 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 200 mm
    1 * 200 mm - 60,40 €
    60,40 €
    1 * 200 mm
       60,40 €