FAM testing fluids C
FAM test fluids C contains a methanolic lower layer for polymer materials.
: According to DIN 51604-3:1984.
- Environmentally Preferable:
Frequently Bought Together
FAM test fluids C contains a methanolic lower layer for polymer materials.