Orders LinkContactUsLinkComponent
Microscopic dissection set
Microscopic dissection set
Microscopic dissecting set, 17 instruments in a wooden case
N° de catalogue HAMMHSO122-00
Fournisseur:   HAMMACHER KARL
Microscopic dissection set
N° de catalogue HAMMHSO122-00
Fournisseur:  HAMMACHER KARL
Microscopic dissection set
Microscopic dissection set
Microscopic dissecting set, 17 instruments in a wooden case
Prix
1 020,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
17 dissection instruments in a wooden case.
Contents:
HSB 715-25 Scalpel
HSB 716-30 Scalpel
HSB 717-35 Scalpel
HSB 718-40 Scalpel
HSB 719-45 Scalpel
HSB 812-18 Cartilage knife
HSC 048-10 Dissecting forceps
HSC 550-10 Dissecting forceps
HSK 257-04 Chisel
HSB 084-11 Dissecting scissors
HSB 006-10 Microscopic scissors
HSO 102-01 Dissecting Needle
HSO 104-03 Dissecting lancet
HSO 095-10 Hook retractor
HSO 092-00 Blow pipe, double
HWP 026-10 Probe
HWP 029-10 Probe
Specifications
  • Description:
    Microscopic dissecting set, 17 instruments in a wooden case
  • Seq:
    04