Order Entry
Belgium
ContactUsLinkComponent
pH/mV/°C meter, handheld, GMH 3511
pH/mV/°C meter, handheld, GMH 3511
Supplier:  Greisinger
Certificates
undefined
pH/mV/°C meter, handheld, GMH 3511
Supplier:  Greisinger

Specifications

  • Catalog No:
  • GRSG605021
  • GRSG604953
  • pH range:
  • 0,00 - 14,00
  • pH resolution:
  • 0,01
  • pH accuracy:
  • ±0,01
  • pH calibration:
  • 2-point calibration with buffer capsules (GPH 4, 7, 10)
  • mV range:
  • −1999...+2000; −1792...+2207 mVH (based on hydrogen system, acc. to DIN 38404)
  • mV resolution:
  • 0,01
  • mV accuracy:
  • ±0.1 % FS (mV bzw. mVH)
  • Temperature range:
  • −5,0...+150 °C
  • Temperature resolution:
  • 0,1 °C
  • Temperature accuracy:
  • ±0,2 °C (−5...+100 °C)
  • Temperature compensation:
  • Automatic (ATC) in the operation mode "pH" if temperature probe is used (0...105 °C); otherwise manual input
  • Display:
  • LCD (two lines, 2×4 digits, 12,4 + 7 mm high)
  • Power supply:
  • 9 V battery, additional socket for external 10,5 - 12 V direct current power supply
  • Connections:
  • BNC, 2× 4 mm banana sockets for Pt1000
  • Outputs:
  • Serial interface (3,5 mm jack); analog output
  • W×D×H:
  • 71×26×142 mm
  • Weight:
  • 0,170 kg

Specifications

Frequently Bought Together