Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, gabapentin, CRM, Supelco®
Organic reference standards, gabapentin, CRM, Supelco®
Catalog # PHR1049-1G
Supplier:  Merck
CAS Number:  
undefined
Organic reference standards, gabapentin, CRM, Supelco®
Catalog # PHR1049-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Standards:
    Reference standard
  • CAS No.:
    60142-96-3
  • Description:
    Gabapentin, pharmaceutical secondary standard
  • Formula:
    C₉H₁₇NO₂
  • Description:
    Gabapentin, pharmaceutical secondary standard

Specifications

About this item

They are pharmaceutical secondary standards for application in quality control, pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Certifications: Accordance with ISO/IEC 17025:2005 (A Class Cert AT-1467), ISO guides 34:2009 (A Class Cert AR-1470).