Organic reference standards, mepyramine maleates, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:52818-63-0
- Formula:C₁₃H₁₄N₂O
- Description:Mepyramine impurity A, N-(4-methoxybenzyl)pyridin-2-amine
Specifications
Frequently Bought Together