Organic reference standards, marbofloxacins, Ph. Eur.
Catalog # EDQMY0000819
Supplier: EDQM
CAS Number:
Specifications
- Pk:10 mg
- Standards:Reference standard
- CAS No.:115550-35-1
- Description:Marbofloxacin, Ph. Eur. standard
- Seq:01
- Formula:C₁₇H₁₉FN₄O₄
- Description:Marbofloxacin