Orders LinkContactUsLinkComponent
Padlocks, brass
Supplier:   Brady
Padlocks, brass
Supplier:  Brady
Certificates
Padlocks, brass
6 Options Available Below
Product Details & Documents
These durable padlocks feature solid brass bodies and shackles that resist corrosion and sparks.
  • 6-pin cylinder offers more unique key cuts for better tamper resistance
  • Locks are protected by coloured vinyl sheaths making it easy to colour-code lockout procedures
  • Engraving on bottom and key
  • Padlocks which are keyed differently are available
Shackle clearance: 20,5 mm
Shackle diameter: 6,5 mm
Shackle height: 21,00 mm
Shackle width: 33,5 mm
 : Lock labels sold separately. Two keys are supplied with each padlock.
Product Family Options
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • Red
    43×17×60 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 158,00 €
    158,00 €
    1 * 12 ST
       158,00 €
  • Blue
    43×17×60 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 158,00 €
    158,00 €
    1 * 12 ST
       158,00 €
  • Green
    43×17×60 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 158,00 €
    158,00 €
    1 * 12 ST
       158,00 €
  • Yellow
    43×17×60 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 158,00 €
    158,00 €
    1 * 12 ST
       158,00 €
  • Grey
    43×17×60 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 158,00 €
    158,00 €
    1 * 12 ST
       158,00 €
  • Black
    43×17×60 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 158,00 €
    158,00 €
    1 * 12 ST
       158,00 €