Orders LinkContactUsLinkComponent
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
Supplier:   Brady
Universal cable lockouts, SafeLex
Supplier:  Brady
Certificates
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
6 Options Available Below
Product Details & Documents
The integrated hasp and cable in these cable lockouts provide a simple way to lock out multiple electrical or valve energy isolation points.
  • Lightweight construction for easy handling, carrying and operation
  • Device comes complete with vinyl coated steel lockout cable attachment
  • Allows up to six safety padlocks for a secure lockout
Product Family Options
Product Information
  • Type
    Availability
    Price
  • With 1 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,10 €
    53,10 €
    1 * 1 ST
       53,10 €
  • With 2 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,70 €
    60,70 €
    1 * 1 ST
       60,70 €
  • With 2 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,70 €
    60,70 €
    1 * 1 ST
       60,70 €
  • With 3 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,30 €
    68,30 €
    1 * 1 ST
       68,30 €
  • With 3 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,30 €
    68,30 €
    1 * 1 ST
       68,30 €
  • With 5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,90 €
    75,90 €
    1 * 1 ST
       75,90 €