Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.01 M (0.01 N) in aqueous solution, Titripur®, Supelco®
Sodium hydroxide 0.01 M (0.01 N) in aqueous solution, Titripur®, Supelco®
Catalog # 1.60309.4003
Supplier:  Merck
CAS Number:  
Sodium hydroxide 0.01 M (0.01 N) in aqueous solution, Titripur®, Supelco®
Catalog # 1.60309.4003
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    PE/alu Titripac® with RFID tag
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Form:
    Liquid
  • Amount of substance:
    0,00995 - 0,01005 mol/l
  • Measurement uncertainity:
    +/- 0,00003 mol/l
  • Traceability:
    NIST SRM

Specification Test Results

About this item

 :
 :40 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548