Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Water
Water, Technical Grade
Water, Technical Grade
:
PanReac AppliChem
Water, Technical Grade
:
PanReac AppliChem
:
7732-18-5
:
:
H₂O
:
MFCD00011332
:
18.02 g/mol
:
100 °C
:
0 °C
:
1.00 kg/l (20 ˚C)
Form:
Liquid
Sterility:
Not tested
Conductivity:
Max. 5,0 μS/cm
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Accessories for RQflex
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
VWR® Rota®, pH indicator paper, 1 to 14
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
: Merck
VWR®, Erlenmeyer Flasks, Narrow Neck, Heavy-Duty
: VWR Collection
Test strips for peroxide, MQuant®, Supelco®
: Merck
pH electrodes, SenTix® PLUS
: WTW