Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, levodropropizines, Ph. Eur.
Organic reference standards, levodropropizines, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, levodropropizines, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    99291-25-5
  • Formula:
    C₁₃H₂₀N₂O₂
  • Description:
    Levodropropizine impurity A, (2R)-3-(4-phenylpiperazin-1-yl)propan-1,2-diol (dextrodropropizine)

Specifications

Frequently Bought Together