Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, hydroxocobalamin, Ph. Eur.
Organic reference standards, hydroxocobalamin, Ph. Eur.
Catalog # EDQMH1428000
Supplier:  EDQM
CAS Number:  
Organic reference standards, hydroxocobalamin, Ph. Eur.
Catalog # EDQMH1428000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    13422-51-0
  • Description:
    Hydroxocobalamin
  • Formula:
    C₆₂H₉₀CoN₁₃O₁₅P
  • Description:
    hydroxocobalamin

Specifications

About this item