Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, glycerols, Ph. Eur.
Organic reference standards, glycerols, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, glycerols, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    4740-78-7
  • Formula:
    C₄H₈O₃
  • Description:
    Glycerol formal

Specifications

Frequently Bought Together