Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, alkali solution (3,0 N)

Buffer, alkali solution (3,0 N)

Alkali buffer solution, 3,0 N, 100 mL
Catalog # HACH2307242
Supplier:   Hach
Buffer, alkali solution (3,0 N)
Catalog # HACH2307242
Supplier:  Hach
CAS Number:  
Buffer, alkali solution (3,0 N)
Buffer, alkali solution (3,0 N)

Alkali buffer solution, 3,0 N, 100 mL

CAS Number:  
Price
28,80 €
Log in for availability
1 * 100 mL
Product Details & Documents

These buffers are used in the determination of calcium and magnesium in brine.

Packaging: Buffers are packed in a 100 ml bottle.

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Alkali buffer solution, 3,0 N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.