Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, ganciclovirs, Ph. Eur.
Organic reference standards, ganciclovirs, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, ganciclovirs, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    82410-32-0
  • Formula:
    C₉H₁₃N₅O₄
  • Description:
    Ganciclovir

Specifications

Frequently Bought Together