Order Entry
Belgium
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cyanine5) [clone: 2D1]
Anti-CD45 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cyanine5) [clone: 2D1]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD45 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cyanine5) [clone: 2D1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 55-9459-T100
  • 55-9459-T025
  • Antigen symbol:
  • CD45
  • CD45
  • Antigen synonyms:
  • Leukocyte Common Antigen, LCA, Ly-5, Hle-1
  • Leukocyte Common Antigen, LCA, Ly-5, Hle-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy5®
  • PE (Phycoerythrin)/Cy5®
  • Clone:
  • 2D1
  • 2D1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Frequently Bought Together