Order Entry
Belgium
ContactUsLinkComponent
Desktop lamps, visionLED
Desktop lamps, visionLED
Catalog # 630-2926
Supplier:  ESCHENBACH OPTIK
undefined
Desktop lamps, visionLED
Catalog # 630-2926
Supplier:  ESCHENBACH OPTIK

Specifications

  • Description:
    Economy visionLED desktop lamp, with 3 colour tones
  • Pk:
    1

Specifications

About this item

High quality desktop lamp offering even, glare-free illumination with high black-white contrast and good colour reproduction.

  • Multi-layer diffuser technology for glare- and hard shadow-free illumination
  • 100% UVA/UVB light blocking
  • Up to 90% blue light damping
  • Colour reproduction index (CRI) of 90
  • LED lifespan: 50000 hours
  • Energy consumption: 10 W
Delivery: Choice of Economy model with 3 colour tones or Comfort model with 5 colour tones and automatic switch off after 2 hours.