Order Entry
Belgium
ContactUsLinkComponent
Multi-element calibration standard 3 without Hg
Multi-element calibration standard 3 without Hg
Catalog # PERCN9301720
Supplier:  PerkinElmer
CAS Number:  
Multi-element calibration standard 3 without Hg
Catalog # PERCN9301720
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Matrix:
    2-5% HNO₃
  • Pk:
    1
  • Description:
    Multi-element calibration standard 3 without Hg

Specifications

About this item

10 µg/ml: Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn