Adjustable handles for brooms and squeegees
: PERFEX
These polymer handles are developed to meet the ergonomic and hygienic needs of controlled environments. They are widely accepted as a superior alternative to fixed length handles based on traditional materials, such as steel, aluminium and wood.
- Comfortable and easy to use
- Durable and lightweight
- Handles will not corrode, oxidise, or rot
- Compatible with gamma and ETO
- Autoclavable up to 121 °C for 30 minutes
These handles are impervious to water, grease, detergents, sanitisers, and solvents. Grasp the upper handle and twist centre grip to adjust the handle, and twist grip back to lock handle securely in place.
With the short connector end, handles can be used with Perfex models no. 2200 and 2300. With a long connector end, the handle is recommended for use with Perfex model no. 2618, 2624, 2518, 2524, 2418 and 3010, as well as squeegee models no. 9518, 9524, and 9530.
Frequently Bought Together