ContactUsLinkComponent
Check valve cartridges, OPTI-MAX®
Check valve cartridges, OPTI-MAX®
Catalog # OPTT10-58-02004
Check valve cartridges, OPTI-MAX®
Catalog # OPTT10-58-02004
Specifications
  • Description:
    Cartridge, standard, with ceramic ball and seat, 3,175 mm OD
  • Material:
    PEEK
  • Seq:
    07
  • Pk:
    2
Specifications
About this item
A single OPTI-MAX® cartridge is interchangeable across multiple pump brands and can function in either the inlet or outlet position.
  • Each ball and seat set is individually matched to ensure a lock and key fit
  • Stainless steel body has high mechanical strength
  • PEEK cartriges are suitable for biocompatible HPLC systems