Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Oasis® WAX

Solid phase extraction cartridges, Oasis® WAX

Cartridge, WAX, 80 Å, 60 µm, 3 ml, 60 mg
Catalog # WATE186002492
Marketsource
Supplier:   WATERS
Solid phase extraction cartridges, Oasis® WAX
Catalog # WATE186002492
Supplier:  WATERS
Marketsource
Solid phase extraction cartridges, Oasis® WAX

Cartridge, WAX, 80 Å, 60 µm, 3 ml, 60 mg

Price
465,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

These Oasis® WAX cartridges contain the WAX sorbent, which is a polymeric reversed phase, weak anion exchange mixed mode sorbent that has been optimised for fast, simple, and highly selective sample preparation of strong acidic compounds.

  • Water wettable sorbent

The 60 µm particle size is recommended when working with viscous samples. These syringe barrel type cartridges are designed for use with vacuum manifolds and automated SPE instruments.

Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    60 µm
  • Pore size:
    80 Å
  • Capacity:
    3 ml
  • Bed weight:
    60 mg
  • Phase:
    WAX