Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tubidity standards for 2100A

Tubidity standards for 2100A

Supplier:   Hach
Tubidity standards for 2100A
Supplier:  Hach
Certificates
Tubidity standards for 2100A
Tubidity standards for 2100A

2 Options Available Below

Product Details & Documents

Stablcal Stabilised Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter.

  • Precise, pre-mixed formazin solutions
  • Based on Formazin Primary Standards
  • Enhanced stability saves time and labour
  • Confidence in results

Documents

Product Family Options

Product Information

  • Turbidity
    Pack type
    Pk Info
    Pk
    Availability
    Price
  • HACH2659105
    StabCal® standard calibration kit for model 2100A
    StabCal® standard calibration kit for model 2100A
    1,0 NTU
    Sealed vials
    4× sealed vial
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 464,00 €
    464,00 €
    1 * 1 KIT
       464,00 €

  • HACH2659110
    StabCal® standard calibration kit for model 2100A
    StabCal® standard calibration kit for model 2100A
    1,0 NTU
    100 ml bottles
    4× 100 ml
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 415,00 €
    415,00 €
    1 * 1 KIT
       415,00 €