Viscosity flow cup
Viscosity cup, 130 g, Lory, 49 mm
Catalog # SIMEVF2199
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Lory viscosity cup designed for quick measurements on location or during production processes.
- Serial number, easy to clean for quick testing
- Brass zinc plate cup with steel leg (height of 170 mm)
- Bottom of this cup provided with a small gap as well as a needle point
The flow time is determined when the product flows out of the small gap and as soon as the needle point appears.
Documents
TQC Company Video
Vidéos
Specifications
- Model:Lory
- Description:Viscosity cup, 130 g
- Cup orifice:49 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



