Order Entry
Belgium
ContactUsLinkComponent
 
 
 
 
Toluene ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
  8608-26
 :  
undefined
 
 
 
Toluene ≥99.5% (by GC), AR® ACS, Macron Fine Chemicals™
  8608-26
 :  

 

  • Pack type:
    Steel drum
  • Pk:
    200 L
  • Seq:
    5549

 

 

  • For Laboratory,Research,or Manufacturing Use:
    ≥ 99.5%
  • ACS - Assay (C₆H₅CH₃) (by GC):
    ≥ 99.5 %
  • Benzene (C₆H₆), For Information On ly:
    ≤10
  • ACS - Color (APHA):
    ≤ 10
  • ACS - Residue after Evaporation:
    ≤ 0.001 %
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Sulfur Compounds (as S):
    ≤ 0.003 %
  • ACS - Water (H₂O)(by KF, coulometric):
    ≤ 0.03 %

 

 

 :
 :92.14 g/mol
 :13,09607
 :110.6 °C (1013 hPa)
 :–95 °C
 :0.867 g/cm³ (20 °C)
 :4 °C
 :Ambient
 :203-625-9
 :1294
 :3,II
 :Management of Change (MOC) category = N
 :05-2114615130-69
 :GHS02
 :GHS08
 :GHS07
 :108-88-3
 :C₆H₅CH₃
 :MFCD00008512