Order Entry
Belgium
ContactUsLinkComponent
Tetrahydrofuran ≥99.0% (by GC, corrected for water content) stabilised, AR® ACS, Macron Fine Chemicals™
Tetrahydrofuran ≥99.0% (by GC, corrected for water content) stabilised, AR® ACS, Macron Fine Chemicals™
Catalog # 8498-26
CAS Number:  
undefined
Tetrahydrofuran ≥99.0% (by GC, corrected for water content) stabilised, AR® ACS, Macron Fine Chemicals™
Catalog # 8498-26
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    400 lb
  • Seq:
    7284
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance (clear, colorless liquid):
    Passes Test
  • ACS - Assay (C₄H₈O) (by GC, corrected for water):
    ≥99.0 %
  • ACS - Color (APHA):
    ≤20
  • ACS - Peroxide (as H₂O₂)(at time of packaging):
    ≤0.015 %
  • ACS - Residue after Evaporation:
    ≤0.03 %
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %

Specification Test Results

About this item

Synonyms: Butylene oxide, THF, Diethylene oxide
 :
 :72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356