Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 50%, CMOS for microelectronic, J.T.Baker®
Corrosive
Sulphuric acid 50%, CMOS for microelectronic, J.T.Baker®
Catalog # 5253-09
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Sulphuric acid 50%, CMOS for microelectronic, J.T.Baker®
Catalog # 5253-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    600 lb
  • Seq:
    7350

Specifications

Specification Test Results

  • For Microelectronic Use:
    49.0 - 51.0 %
  • Assay (H₂SO₄):
    49.0 - 51.0 %
  • Color (APHA):
    ≤10
  • Residue after Ignition:
    ≤3 ppm
  • Chloride (Cl):
    ≤1 ppm
  • Nitrate (NO₃):
    ≤0.2 ppm
  • Phosphate (PO₄):
    ≤0.5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤0.100 ppm
  • Arsenic and Antimony (as As):
    ≤0.005 ppm
  • Trace Impurities - Barium (Ba):
    ≤1.000 ppm
  • Trace Impurities - Boron (B):
    ≤0.010 ppm
  • Trace Impurities - Cadmium (Cd):
    ≤0.500 ppm
  • Trace Impurities - Calcium (Ca):
    ≤1 ppm
  • Trace Impurities - Chromium (Cr):
    ≤0.500 ppm
  • Trace Impurities - Cobalt (Co):
    ≤0.500 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.010 ppm
  • Trace Impurities - Gallium (Ga):
    ≤0.050 ppm
  • Trace Impurities - Germanium (Ge):
    ≤0.500 ppm
  • Trace Impurities - Gold (Au):
    ≤0.500 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.2 ppm
  • Trace Impurities - Lead (Pb):
    ≤0.400 ppm
  • Trace Impurities - Lithium (Li):
    ≤1.000 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤1.000 ppm
  • Trace Impurities - Manganese (Mn):
    ≤0.500 ppm
  • Trace Impurities - Nickel (Ni):
    ≤0.1 ppm
  • Trace Impurities - Potassium (K):
    ≤1.000 ppm
  • Trace Impurities - Silicon (Si):
    ≤0.500 ppm
  • Trace Impurities - Silver (Ag):
    ≤0.500 ppm
  • Trace Impurities - Sodium (Na):
    ≤1.000 ppm
  • Trace Impurities - Strontium (Sr):
    ≤0.500 ppm
  • Trace Impurities - Tin (Sn):
    ≤0.500 ppm
  • Trace Impurities - Zinc (Zn):
    ≤0.500 ppm

Specification Test Results

About this item

 :
 :98.08 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:1.1…1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589