Order Entry
Belgium
ContactUsLinkComponent
 
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) ACS, USP, J.T.Baker®
Catalogus # 3808-65
Leverancier:  Avantor
CAS-nummer:  
undefined
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) ACS, USP, J.T.Baker®
Catalogus # 3808-65
Leverancier:  Avantor
CAS-nummer:  

Specificaties

  • Pack type:
    HDPE drum, Flowmor™ package
  • Pk:
    100 kg
  • Seq:
    4553
  • Environmentally Preferable:

Specificaties

Specificatie testresultaten

  • GMP Manufactured Product:
    98.0 - 103.0 %
  • Meets ACS Reagent Chemical Requirements:
    Passes Test
  • Meets U.S.P Requirements:
    = 8 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    = 0.014 %
  • Bulk Pharmaceutical Chemical:
    = 0.002 %
  • USP - Assay (anhydrous basis):
    98.0 - 103.0 %
  • USP - Aluminum, Calcium, and Related Elements:
    Passes Test
  • USP - Arsenic (As):
    ≤8 ppm
  • USP - Chloride (Cl):
    ≤0.014 %
  • USP - Identification:
    Passes Test
  • USP - Insoluble Substances:
    ≤0.2 %
  • USP - pH (1 in 20):
    4.1 - 4.5
  • USP - Sulfate (SO₄):
    ≤0.15 %
  • USP - Water (H₂O):
    10.0 - 15.0 %
  • Assay (NaH₂PO₄·H₂O):
    98.0 - 102.0 %
  • pH of 5% Solution at 25°C:
    4.1 - 4.5
  • Insoluble Matter:
    ≤0.01 %
  • Chloride (Cl):
    ≤5 ppm
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.005 %
  • Potassium (K):
    ≤0.01 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %

Specificatie testresultaten

Over dit artikel

Synoniemen: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
 :
 :137.99 g/mol
Merck index:13,08734
Smelten Pt:100 °C
Dichtheid:2.04 g/cm³
Temperatuur bij opslag:Ambient
EINECS:231-449-2
BEREIKEN:01-2119489796-13
CAS-nummer:10049-21-5
Formule:NaH₂PO₄·H₂O
MDL-nummer:MFCD00149208