
Sodium metabisulphite NF, J.T.Baker®
3550-05
: Avantor
:
- Pk:2,5 kg
- Seq:4076
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Limit of Chloride:≤0.05 %
- NF - Iron (Fe):≤0.002 %
- NF - Limit of Thiosulfate:≤0.05 %
- NF - Assay (SO₂):65.0 - 67.4 %
:
:190.11 g/mol
:1.48 g/cm³
:GHS05
:7681-57-4
:Na₂S₂O₅
:MFCD00167602