Order Entry
Belgium
ContactUsLinkComponent
Sodium hypochlorite ≥10% available chlorine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sodium hypochlorite ≥10% available chlorine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Sodium hypochlorite ≥10% available chlorine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Available Chlorine:
    min. 10 %
  • Calcium (Ca):
    max. 0.001 %
  • Phosphate (as PO4):
    max. 0.0005 %

Specification Test Results

Related Information

Frequently Bought Together