Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide concentrate for dilution to 0.5 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide concentrate for dilution to 0.5 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4691.
Supplier:  Avantor
CAS Number:  
undefined
Sodium hydroxide concentrate for dilution to 0.5 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4691.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    210 mL
  • Seq:
    2545

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • Normality (by titrimetry):
    Passes Test
  • (Makes 0.5N solution after dilution to 1000 mL):
  • Low in Carbonate:

Specification Test Results

About this item

 :
 :40 g/mol
UN:1823
ADR:8,II
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548