Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide, J.T.Baker®

Sodium hydroxide, J.T.Baker®

1.000 L
Catalog # 2632-40
Supplier:   Avantor
Sodium hydroxide, J.T.Baker®
Catalog # 2632-40
Supplier:  Avantor
CAS Number:  1310-73-2
Sodium hydroxide, J.T.Baker®

1.000 L

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 1.000 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • UN:1823
  • ADR:8,II
Specifications

Specification Test Results

  • Assay (NaOH):
    16.0 - 18.0 %
  • Appearance (clear, colorless liquid):
    passes test
  • Identification (Na):
    passes test
  • Sodium Carbonate (Na2CO3):
    max. 0.6 %
  • pH at 25°C:
    min. 11.0
  • Trace Impurities (in ppm)Chlorides (as Cl):
    max. 50
  • Trace Impurities (in ppm) Iron (Fe):
    max. 10
  • Trace Impurities (in ppm) Heavy Metals (as Pb):
    max. 20
  • Trace Impurities (in ppm) Sulfate (SO4):
    max. 50

Specifications

  • Pk:
    1.000 L

Specifications