Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 50% (by acidimetry), BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Corrosive
Sodium hydroxide 50% (by acidimetry), BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Catalog # 3735-07
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Sodium hydroxide 50% (by acidimetry), BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Catalog # 3735-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    65 lb
  • Seq:
    7149

Specifications

Specification Test Results

  • Assay (NaOH) (by acidimetry):
    49.5 - 51.5 %
  • Color (APHA):
    ≤10
  • Insoluble Matter:
    ≤30 ppm
  • Sodium Carbonate (Na₂CO₃):
    ≤0.05 %
  • Ammonium Hydroxide Precipitate:
    ≤0.007 %
  • Potassium (K):
    ≤0.005 %
  • Chloride (Cl):
    ≤20 ppm
  • Nitrogen Compounds (as N):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤5 ppm
  • Sulfate (SO₄):
    ≤10 ppm
  • Heavy Metals (as Ag):
    ≤10 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - Nickel (Ni):
    ≤3.000 ppm

Specification Test Results

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548