Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 49.5 - 51.5% (by acidimetry) in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Corrosive
Sodium hydroxide 49.5 - 51.5% (by acidimetry) in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Sodium hydroxide 49.5 - 51.5% (by acidimetry) in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (NaOH) (by acidimetry):
    49.5 - 51.5 %
  • Color (APHA):
    ≤10
  • Insoluble Matter:
    ≤30 ppm
  • Sodium Carbonate (Na₂CO₃):
    ≤0.05 %
  • Ammonium Hydroxide Precipitate:
    ≤0.005 %
  • Chloride (Cl):
    ≤20 ppm
  • Nitrogen Compounds (as N):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤5 ppm
  • ACS - Sulfate (SO₄):
    ≤5 ppm
  • Trace Impurities - Chromium (Cr):
    ≤5.0 ppm
  • Trace Impurities - Copper (Cu):
    ≤1 ppm
  • Heavy Metals (as Ag):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - Nickel (Ni):
    ≤3.000 ppm
  • Trace Impurities - Potassium (K):
    ≤50 ppm
  • Trace Impurities - Zinc (Zn):
    ≤5.000 ppm

Specification Test Results

Related Information

Frequently Bought Together