Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Sodium hydroxide 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    50 - 52%
  • Store above 16°C (60°F) to prevent freezing:
    ≤5 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤1 ppm
  • Assay (NaOH):
    50 - 52 %
  • Chloride (Cl):
    ≤ 30 ppm
  • Heavy Metals (as Pb):
    ≤ 1 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 2 ppm
  • Sodium Carbonate (Na₂CO₃):
    ≤ 3.0 %

Specification Test Results

Related Information

Frequently Bought Together