Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Sodium hydroxide 5.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    4.9 - 5.1
  • Protect from air to avoid absorption of carbon dioxide.:
    ≤5 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤1 ppm
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Normality:
    4.9 - 5.1
  • Chloride (Cl):
    ≤10 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Iron (Fe):
    ≤2 ppm
  • Endotoxin Concentration (EU/mL:
    ≤2

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together