Order Entry
Belgium
ContactUsLinkComponent
Sodium formate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium formate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium formate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • ACS - Assay (HCOONa):
    ≥99.0 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.001 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together