Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride ≥99.0% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Sodium chloride ≥99.0% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Sodium chloride ≥99.0% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    5.0 - 9.0
  • Assay (NaCl) (by Ag titrn):
    ≥99.0 %
  • pH of 5% Solution at 25°C:
    5.0 - 9.0
  • Insoluble Matter:
    ≤0.005 %
  • Iodide (I):
    ≤0.002 %
  • Bromide (Br):
    ≤0.01 %
  • Chlorate and Nitrate (as NO₃):
    ≤0.003 %
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • Sulfate (SO₄):
    ≤0.004 %
  • Barium (Ba):
    Passes Test
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Iron (Fe):
    ≤2 ppm
  • Calcium (Ca):
    ≤0.002 %
  • Magnesium (Mg):
    ≤0.001 %
  • Potassium (K):
    ≤0.005 %

Specification Test Results

Related Information

Frequently Bought Together