Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Irritant
Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Irritant
Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (NaCl) (by Ag titrn):
    ≥99.0 %
  • ACS - pH of 5% Solution at 25°C:
    5.0 - 9.0
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iodide (I):
    ≤0.002 %
  • ACS - Bromide (Br):
    ≤0.01 %
  • ACS - Chlorate and Nitrate (as NO₃):
    ≤0.003 %
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • ACS - Sulfate (SO₄):
    ≤0.004 %
  • ACS - Barium (Ba):
    Passes Test
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤2 ppm
  • ACS - Calcium (Ca):
    ≤0.002 %
  • ACS - Magnesium (Mg):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.005 %

Specification Test Results

Frequently Bought Together