Sodium carbonate monohydrate ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Sodium carbonate hydrous, Soda monohydrate
CAS Number:5968-11-6
:
Formula:Na₂CO₃·1H₂O
MDL Number:MFCD00149177
Molecular Weight:124.00 g/mol
Melting Pt:107.85
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.5 %
- ACS - Assay (Na₂CO₃·H₂O):≥99.5 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Loss on Drying at 150°C:13.0 - 15.0 %
- ACS - Chloride (Cl):≤0.001 %
- ACS - Calcium (Ca):≤0.03 %
- ACS - Silica (SiO₂):≤0.005 %
- ACS - Sulfur Compounds (as SO₄):≤0.004 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Potassium (K):≤0.005 %
- Phosphate (PO₄):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
Frequently Bought Together