
Sodium carbonate, anhydrous ≥99.5% (dried basis), powder, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:497-19-8
:
:Na₂CO₃
:MFCD00003494
:105.99 g/mol
:14,08596
:1600 °C (1013 hPa)
:854 °C
:2.53 g/cm³ (20 °C)
:Management of Change (MOC) category = N
:Ambient
:207-838-8
:01-2119485498-19
:GHS07
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Assay (Na₂CO₃) (dried basis)(by acidimetry):≥99.5 %
- Insoluble Matter:≤0.01 %
- Loss on Heating at 285°C:≤1.0 %
- Chloride (Cl):≤0.001 %
- Phosphate (PO₄):≤0.001 %
- Silica (SiO₂):≤0.005 %
- Sulfur Compounds (as SO₄):≤0.003 %
- Calcium (Ca):≤0.03 %
- Magnesium (Mg):≤0.005 %
- Potassium (K):≤0.005 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Meets Reagent Specifications for testing USP/NF monographs:—
Frequently Bought Together