Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Irritant
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Catalog # 3642-26
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Catalog # 3642-26
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE drum, open head
  • Pk:
    200 lb
  • Seq:
    7197

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Water (H₂O):
    ≤0.5 %
  • NF - Assay (Na₂CO₃) (anhydrous basis):
    99.5 - 100.5 %
  • ACS - Assay (Na₂CO₃) (dried basis)(by acidimetry):
    ≥99.5 %
  • ACS - Calcium (Ca):
    ≤0.03 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Loss on Heating at 285°C:
    ≤1.0 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Silica (SiO₂):
    ≤0.005 %
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.003 %

Specification Test Results

About this item

 :
 :105.99 g/mol
 :GHS07
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494